Storage Vessels, #15 O-Ring Joint, Chemscience, SATI International Scientific

Supplier: CHEM SCIENCE INC CA
CS-S0490025 CS-S0490050 CS-S0490100 CS-S0490350 CS-S0520050 CS-S0520100 CS-S0520250 CS-S0520500 CS-S0521000
CA10203-034CS 276.01 CAD
CA10203-034 CA10203-036 CA10203-038 CA10203-040 CA10203-042 CA10203-044 CA10203-046 CA10203-048 CA10203-050
Storage Vessels, #15 O-Ring Joint, Chemscience, SATI International Scientific
Reaction Vessels Reaction Vessels, Flask Shape
Storage vessel with a #15 O-ring joint on the side arm and a high vacuum valve to control gas or liquid flow into the vessel.

Delivery information: Supplied complete with one #116 Viton® O-ring.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR