Sublimators, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0732510 CS-S0734040 CS-S0735060
CA10203-004CS 636.79 CAD
CA10203-004 CA10203-006 CA10203-008
Sublimators, Chemscience
Condensers
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed.
  • Sublimate collects on the lower end of the condenser which has a flat bottom
  • Condenser is constructed using a grease-free, threaded valve
Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery.
Delivery information: Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring and stainless steel pinch clamp.