Stopcocks, PTFE, Four Way, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0421530 CS-S0341750 CS-S0342565 CS-S0701240 CS-S0701754 CS-S0701756 CS-S0711240 CS-S0711754 CS-S0711756 CS-S0721240 CS-S0721754 CS-S0721756
CA10202-978CS 326.87 CAD
CA10202-978 CA10202-980 CA10202-982 CA10202-986 CA10202-988 CA10202-990 CA10202-992 CA10202-994 CA10202-996 CA10202-998 CA10203-000 CA10203-002
Stopcocks, PTFE, Four Way, Chemscience
Stopcocks
A 1:5 taper PTFE plug having double 'V', 90° bores to connect opposite sets of adjacent rams.
Configuration of stopcock allows it to be used in a system to obtain reversal of flow.
Order Now
SPECIFICATIONS
Environmentally Preferable