Chemscience Solvents, Distilling Heat, SATI International Scientific
Supplier: SATI International Scientific
CS-S0254000 CS-S0259500 CS-S0259250 CS-S025242L CS-S0254250 CS-S0251250 CS-S0254500 CS-S0251500
CA10202-916CS 1045.12 CAD
CA10202-916 CA10202-918 CA10202-920 CA10202-922 CA10202-912 CA10202-924 CA10202-914 CA10202-926
Chemscience Solvents, Distilling Heat, SATI International Scientific
Distillation Components Distillation Heads
Upper sampling port has a 4 mm PTFE stopcock with a 14/20 outer joint for use with septa.
Lower drain is centered to allow total drainage of storage head.