Palladium(II) nitrate 10.0 ± 0.2 g/L in nitric acid 15% matrix modifier solution, for graphite furnace AAS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.07289.0050 1.07289.0050
CA1.07289.0050CS 26286.32 CAD
CA1.07289.0050 EM1.07289.0050
Palladium(II) nitrate 10.0 ± 0.2 g/L in nitric acid 15% matrix modifier solution, for graphite furnace AAS, Supelco®
Palladium (II) nitrate

Palladium matrix modifier for graphite furnace AAS.

Formula: N₂O₆Pd
MW: 230.43 g/mol
Density: 1.11 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Assay (palladium) 9.8 - 10.2 g/l
Ag (Silver) ≤ 0.000020 g/l
As (Arsenic) ≤ 0.000010 g/l
Cd (Cadmium) ≤ 0.000001 g/l
Pb (Lead) ≤ 0.000010 g/l
Se (Selenium) ≤ 0.00010 g/l
Tl (Thallium) ≤ 0.000020 g/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR