WHEATON® Safety Coated Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
220735 220736 220935 220936
470200-646CS 765.85 CAD
470200-646 470200-648 470200-650 470200-652
WHEATON® Safety Coated Bottles, DWK Life Sciences
Bottles Glass Bottles
These safety coated bottles were made from clear or amber soda-lime glass that conforms to USP Type III requirements.
  • Available with or without caps
  • 4, 8, 16 and 32 oz. Boston Round style
  • 80 oz. and 4 L with jug handle
  • Plastisol coating for safety