Solvent Stills, High Vacuum Valves, Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-S0264250 CS-S0264500 CS-S0274125
CA10202-904CS 1465.59 CAD
CA10202-904 CA10202-906 CA10202-910
Solvent Stills, High Vacuum Valves, Chemscience, SATI International Scientific
Distillation Components Distillation Heads
These solvent still high vacuum valves have PTFE covers on the shaft and tip.
  • Syringe port has a 4 mm PTFE stopcock and a 14/20 outer joint
  • Delivery port has a 14/20 inner joint
For greater solvent/chemical resistance, the two 0 to 4 mm high vacuum valves have PTFE covers over the two Viton® O-rings on the shaft and one integral PTFE O-rings on the tip, without exposing any material other than PTFE to any solvents. Syringe port has a 4 mm PTFE stopcock and a 14/20 outer joint. Delivery port has a 14/20 inner joint.