Solvent Stills, Repurification (Grease-Free), Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-S0134250 CS-S0134500
CA10202-892CS 618.61 CAD
CA10202-892 CA10202-894
Solvent Stills, Repurification (Grease-Free), Chemscience, SATI International Scientific
Distillation Components Distillation Heads
Access port has threaded 14/10 joint to accept a PTFE-lined silicone septum with a compression cap (supplied).
Threaded 14/10 also accept regular 14/20 glass or PTFE stopper.
Ordering information: Condenser and flasks are not supplied. Stopper not supplied.
Delivery information: Head only. Supplied with compression cap.