Pressure Vessels, Heavy Wall, Round Bottom, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-P0170001 CS-P0170002 CS-P0170003 CS-P0170004 CS-P0170005 CS-P0170006
CA10202-880CS 161.26 CAD
CA10202-880 CA10202-882 CA10202-884 CA10202-886 CA10202-888 CA10202-890
Pressure Vessels, Heavy Wall, Round Bottom, Chemscience
Reaction Vessels Reaction Vessels, Flask Shape
These vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal.
  • Unique design allows hand tightening and loosening for easy access into the vessel
  • O-ring is positioned on the bushing above the thread for improved sealing
The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime.
Delivery information: Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring.