Pressure Vessels, Heavy wall, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-P0160001 CS-P0160002 CS-P0160003 CS-P0160004 CS-P0160005 CS-P0160006
CA10202-868CS 110.77 CAD
CA10202-868 CA10202-870 CA10202-872 CA10202-874 CA10202-876 CA10202-878
Pressure Vessels, Heavy wall, Chemscience
Reaction Vessels Reaction Vessels, Cylindrical Shape
These vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal.
  • Unique design allows hand tightening and loosening for easy access into the vessel
  • O-ring is positioned on the bushing above the thread for improved sealing
The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime.
Delivery information: Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring.