Peptide Synthesis Vessels, Solid Phase, with Lower Joint and T-Bore PTFE Plug, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-P0150025C CS-P0150025M CS-P0150050C CS-P0150050M CS-P0150100C CS-P0150100M CS-P0150250C CS-P0150250M CS-P0150500C CS-P0150500M
CA10202-848CS 383.71 CAD
CA10202-848 CA10202-850 CA10202-852 CA10202-854 CA10202-856 CA10202-858 CA10202-860 CA10202-862 CA10202-864 CA10202-866
Peptide Synthesis Vessels, Solid Phase, with Lower Joint and T-Bore PTFE Plug, Chemscience
Reaction Vessels Reaction Vessels, Cylindrical Shape
These solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation.
  • 8 mm hose connection for vacuum
  • Stopcock has an 8 mm O.D. side arm for inert gas introduction
The 24/40 inner drip joint can be assembled with other standard glassware.
Delivery information: Vessels are supplied with PTFE-faced silicone lined black Nylon® caps.