Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-P0140010C CS-P0140010M CS-P0140025C CS-P0140025M CS-P0140050C CS-P0140050M CS-P0140100C CS-P0140100M
CA10202-832CS 369.64 CAD
CA10202-832 CA10202-834 CA10202-836 CA10202-838 CA10202-840 CA10202-842 CA10202-844 CA10202-846
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Reaction Vessels Reaction Vessels, Cylindrical Shape
With a fritted disc of coarse or medium porosity to support resin.
  • Top of the vessel has GL thread and a PTFE lined screw cap
  • PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas.
Order Now
SPECIFICATIONS
Environmentally Preferable