Kimtech™ A6 Breathable Liquid and Particle Protection Coveralls, Kimberly-Clark Professional
Supplier: Microflex
47681 47682 47683 47684 47685 47686 47687
CA76376-400CS 513.53 CAD
CA76376-400 CA76376-402 CA76376-404 CA76376-406 CA76376-408 CA76376-410 CA76376-412
Kimtech™ A6 Breathable Liquid and Particle Protection Coveralls, Kimberly-Clark Professional
Disposable Garments
Trust Kimtech™ A6 Breathable Liquid and Particle Protection Coveralls when a high-value combination of quality, comfort, and enhanced chemical and microbiological protection are required.
  • Garments feature hood, thumb loops, and elastic cuffs
  • High-performance laminated film fabric for the hood, front panel, arms, and legs
  • Ultra-lightweight breathable SMS for the back panel
  • Origami-folded to facilitate and reduce gowning time; arms telescope-folded, legs telescope folded with built-in snaps
  • Suitable for IOS 7 and 8 / Grade C and D cleanrooms
  • Low lint fabric,
More Product Information