Peptide Synthesis Vessels, Solid Phase, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-P0120010C CS-P0120010M CS-P0120025C CS-P0120025M CS-P0120050C CS-P0120050M CS-P0120100C CS-P0120100M CS-P0120250M
CA10202-796CS 252.19 CAD
CA10202-796 CA10202-798 CA10202-800 CA10202-802 CA10202-804 CA10202-806 CA10202-808 CA10202-810 CA10202-812
Peptide Synthesis Vessels, Solid Phase, Chemscience
Reaction Vessels Reaction Vessels, Cylindrical Shape
With a fritted disc of coarse or medium porosity to support resin.
  • Top of the vessel has GL thread and PTFE lined screw cap
PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.