Vacuum Manifolds, Single Body, Chemscience, SATI International Scientific
Supplier: SATI International Scientific
CS-M0221922 CS-M0322440 CS-M0221420 CS-M0321420 CS-M0222440 CS-M0321922
CA10202-718CS 1401.18 CAD
CA10202-718 CA10204-488 CA10202-720 CA10202-712 CA10202-714 CA10202-716
Vacuum Manifolds, Single Body, Chemscience, SATI International Scientific
Manifolds
These manifolds have four high vacuum solid glass stopcocks of 4 mm bore and inner joints.
  • T-bore high vacuum stopcock at the inlet end of the main tube
  • Main tube has 22 mm O.D.
All glass stopcocks have constant pressure retention devices.