KIMBLE® Atmospheric Environmental Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
653200-2000 653210-2000
KT653200-2000CS 1025.95 CAD
KT653200-2000 KT653210-2000
KIMBLE® Atmospheric Environmental Bottles, DWK Life Sciences
Bottles Gas Washing Bottles
The design of the atmospheric environmental bottle is recommended by the Atmospheric Environment Service (AES).
  • KimCote® plastic safety coating
  • Borosilicate glass high vacuum plug valve stems instead of PTFE stems, eliminating the chance of outgassing
  • FKM valve stem o-rings that are easily changed
  • Inlet stems ground to ¹/₂" Ext.Ø
  • Manufactured from 33 expansion, low extractable borosilicate glass
Available in either a single or double valve configuration.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.