KIMBLE® Reaction Flasks, Spherical, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
612250-5000
KT612250-5000EA 928.35 CAD
KT612250-5000
KIMBLE® Reaction Flasks, Spherical, DWK Life Sciences
Reaction Vessels Reaction Vessels, Flask Shape
Spherical reaction flasks with a flat ground flange.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Clamp 10135-804 is available as an accessory for the 4" flasks.