KIMBLE® Reaction Flasks, Cylindrical, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
612000-0500 612000-1000 612000-2000 612000-3000
KT612000-0500EA 574.52 CAD
KT612000-0500 KT612000-1000 KT612000-1500 KT612000-2000 KT612000-3000
KIMBLE® Reaction Flasks, Cylindrical, DWK Life Sciences
Reaction Vessels Reaction Vessels, Flask Shape
Cylindrical reaction flasks with a flat ground flange and a ³/₁₆" wall construction.
  • Manufactured from 33 expansion, low extractable borosilicate glass
This 4" flasks accept PTFE blade KT789030-0022 and clamp 10135-804.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.