KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
609400-0224 609400-0624
KT609400-0624EA 702.35 CAD
KT609400-0624 KT609400-0224
KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Flasks Distillation Flasks
Indented three-neck round bottom flasks with standard taper outer joints, all vertical.
  • Heavy wall
  • Indentations enhance agitation when stirring
  • Manufactured from 33 expansion, low extractable borosilicate glass
This flask is also referred to as a Morton flask.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.