KIMBLE® Recovery Flasks with Square Joint Bead, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
608675-4124 608675-4224 608675-4624 608675-4824 608675-6129 608675-6229 608675-6729 608675-4724
KT608675-4124EA 114.01 CAD
KT608675-4124 KT608675-4224 KT608675-4624 KT608675-4824 KT608675-6129 KT608675-6229 KT608675-6329 KT608675-6629 KT608675-6729 KT608675-6829 KT608675-4324 KT608675-4724
KIMBLE® Recovery Flasks with Square Joint Bead, DWK Life Sciences
Flasks Rotary Evaporator Flasks
Single neck flask with a standard taper outer joint designed for easy recovery of reaction products.
  • Square bead joints provide superior clamping to standard joint beads
  • Total contents are accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Manufactured from 33 expansion, low extractable borosilicate glass
Even wall thickness and minimum runout make these flasks ideal for use with rotary evaporators.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.