KIMBLE® Self Washing Top Vapor Tube Traps , DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
570300-0114 570300-0124 570300-2524
KT570300-0114EA 460.17 CAD
KT570300-0114 KT570300-0124 KT570300-2524
KIMBLE® Self Washing Top Vapor Tube Traps , DWK Life Sciences
Vacuum Traps
Kimble® self washing top vapor tube traps with square joint bead, [ST] joints top (outer) and [ST] joints bottom (inner).
  • Tapered sidewalls permit continuous washing with condensed solvent while preventing vapor cooling points along the path of the condenser
  • Upper stem has four large holes that allow solvent vapor to flow freely and help to make this portion of the trap easy to clean
  • Increased volume of the trap prevents 'bumping' and keeps solvent out of the vapor tube
  • Manufactured from 33 expansion, low extractable borosilicate glass
This trap is designed to eliminate hold-up common to the round style of rotary evaporator traps.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.