KIMBLE® Bottom Vapor Tube Traps with Drain Holes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
570265-0114 570265-2514 570265-2524 570265-5024
KT570265-0114CS 505.23 CAD
KT570265-0114 KT570265-2514 KT570265-2524 KT570265-5024
KIMBLE® Bottom Vapor Tube Traps with Drain Holes, DWK Life Sciences
Vacuum Traps
These bottom inner vapor tube features two, 4 mm drain holes at its base placed 180° to each other and 90° to the upper holes, allowing faster drainage of trap contents back into the evaporation flask
  • Modified joint on the KIMBLE® bottom vapor tube traps with drain holes features a square bead for better sealing to rotary evaporator equipment
  • Manufactured from 33 expansion, low extractable borosilicate glass
Use where foaming is likely to occur and be drawn into the condenser.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438More Product Information