KIMBLE® Bottom Vapor Tube Traps, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
570260-2524 570260-2514
KT570260-2514EA 428.21 CAD
KT570260-2514 KT570260-2524
KIMBLE® Bottom Vapor Tube Traps, DWK Life Sciences
Vacuum Traps
This trap is designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping.
  • No drain holes assure that dry residue does not get re-wetted
  • Manufactured from 33 expansion, low extractable borosilicate glass
The modified joint on the KIMBLE® bottom vapor tube traps features a square bead for better sealing to rotary evaporator equipment.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.