KIMBLE® Top Vapor Tube Traps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
570210-0124 570210-2519 570210-0114 570210-0119 570210-2514 570210-2524 570210-5014 570210-5024
KT570210-0124EA 448.88 CAD
KT570210-0124 KT570210-2519 KT570210-0114 KT570210-0119 KT570210-2514 KT570210-2524 KT570210-5014 KT570210-5024
KIMBLE® Top Vapor Tube Traps, DWK Life Sciences
Vacuum Traps
The design of this trap provides for expanded volume for organic mixtures that have a tendency to foam excessively.
  • Inner vapor tube is ring-sealed at the top of the flask to prevent entry into the condenser
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.