KIMBLE® Cyanide Distillation Apparatus, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
479110-0000
KT479110-0000CS 1734.37 CAD
KT479110-0000
KIMBLE® Cyanide Distillation Apparatus, DWK Life Sciences
Distillation Components Distillation Sets
These 19/38 apparatus for quantitative analysis of various industrial waste waters, saline and reagent grade water.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Cyanide as hydrocyanic acid (HCN) is released from cyanide complexes by means of a reflux-distillation and absorbed in a scrubber containing a sodium hydroxide solution. The cyanide ion in the absorbing solution is then determined colorimetrically or by volumetric titration.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.