KIMBLE® Coil-Type Reflux Condensers with Two Upper Hose Barbs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
457000-0125 457000-0175 457000-0225 283010-0000
KT457000-0125EA 906.31 CAD
KT457000-0125 KT457000-0175 KT457000-0225 10000-506
KIMBLE® Coil-Type Reflux Condensers with Two Upper Hose Barbs, DWK Life Sciences
Condensers
Tightly wound coil provides enough surface area to condense high vapor pressure solvents like hexane.
  • With an internal, coil-type cold finger
  • Two hose connectors at the top for water inlet and outlet
  • Ref: ASTM Method D94, D95
  • Manufactured from 33 expansion, low extractable borosilicate glass
Standard taper outer joint at the top, standard taper inner drip joint at the bottom. KT283000-0000 does not have a drip joint.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.