KIMBLE® Coil-Type Reflux Condensers with Upper and Lower Hose Barbs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
456900-3024 456900-2024
KT456900-3024EA 900.75 CAD
KT456900-3024 KT456900-2024
KIMBLE® Coil-Type Reflux Condensers with Upper and Lower Hose Barbs, DWK Life Sciences
Condensers
Reflux condenser has efficient non-flooding design.
  • Full length 24/40 joints
  • Manufactured from 33 expansion, low extractable borosilicate glass
Reflux condenser with a water-cooled inner coil sealed to top and bottom hose connectors.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.