KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
296500-0100 617000-0429 617000-0629 617000-0745 617000-0824 617000-0829 617000-0845 617000-1045 617000-1124 617000-1145 296500-0005 296500-0010 296500-0025 296500-0050 296510-0125 617000-0424 617000-0624 617000-0724 617000-0729 617000-0834 296510-0025 296510-0250
KT296500-0100CS 91 CAD
KT296500-0100 KT617000-0429 KT617000-0629 KT617000-0645 KT617000-0745 KT617000-0824 KT617000-0829 KT617000-0845 KT617000-1024 KT617000-1045 KT617000-1124 KT617000-1145 KT296500-0005 KT296500-0010 KT296500-0015 KT296500-0025 KT296500-0050 KT296510-0125 KT617000-0124 KT617000-0424 KT617000-0624 KT617000-0724 KT617000-0729 KT617000-0834 10000-510 10000-512 10000-514 10000-516
KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Flasks Erlenmeyer Flasks
Single neck flask with a standard taper outer joint.
  • White ceramic enamel marking spot
  • Manufactured from 33 expansion, low extractable borosilicate glass
Tops are reinforced and tooled with a rounded finish, containing more glass to give them maximum mechanical strength. Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.