KIMBLE® Distilling Flask, Pear Shaped with Side Tubulation, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
294500-0010
KT294500-0010EA 136.04 CAD
KT294500-0010
KIMBLE® Distilling Flask, Pear Shaped with Side Tubulation, DWK Life Sciences
Flasks Distillation Flasks
Pear-shaped flask with a standard taper outer joint and 7 mm ID side tabulation.
  • Heavy wall
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.