Anti-TNR16 Mouse Monoclonal Antibody [clone: 8J2]

Supplier: Biosensis
M-1818-100
76334-026EA 556.16 CAD
76334-026
Anti-TNR16 Mouse Monoclonal Antibody [clone: 8J2]
Antibodies
p75NTR (CD271) was originally discovered as a low affinity nerve growth factor receptor (NGFR). Later it was found that it was the receptor for all neurotrophins, including NGF, BDNF, NT3 and NT4/5. It mediates signals of neurotrophins for neuronal survival, apoptosis, neurite outgrowth and synaptic plasticity. Recently, it has been revealed that p75NTR not only acts as the receptor for neurotrophins but also the receptor for many other pathological ligands such as prions, rabies virus and amyloid beta. p75NTR also acts as a co-receptor for NOGO which mediates inhibitory signals of myelin associated protein. p75NTR is highly expressed in a number of non-neuronal and neuronal cells including motor neurons during development and also in damaged neurons. Recent research proposes the extracellular domain of p75NTR as a biomarker for monitoring the progression of motor neuron disease (MND), also known as Amyotrophic Lateral Sclerosis (ALS) or Lou Gehrig's Disease. SUBUNIT: Homodimer; disulfide-linked. Interacts with p75NTR-associated cell death executor. Interacts with NGFRAP1/BEX3.

This antibody is specific for NGFR/p75NTR as demonstrated by western blotting and immunprecipitation. The antibody recognizes extracellular p75NTR under non-reducing conditions.

Applications: Flow Cytometry: 5-20 ug/mL.
Western Blotting: 0.5-2.0 ug/mL, non-reducing conditions only (no DTT or beta-mercaptoethanol).
Immunoprecipitation: lysate dependent. 10 ug per 200-500 ug total protein.
Immunopanning: 1-5 ug/mL.
Immunocytochemistry: 1-5 ug/mL. Staining is strongest in non-fixed cells, light fixation is tolerable.
Immunohistochemistry: fresh, acetone fixed sections only, epitope is fixation sensitive. Not suitable in formalin-fixed, paraffin (FFPE) embedded tissues.
Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNR16
Clonality: Monoclonal
Clone: 8J2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, mouse and rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR