Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7060030AM 7060060AM 7060125AM 7060250AM 7060500AM 7061000AM
76360-376PK 59.78 CAD
76360-376 76360-378 76360-380 76360-382 76360-384 76360-386
Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, High Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These narrow mouth bottles improve pouring of liquid contents making them perfect for storing and shipping liquids. Amber bottles reduce UV light transmission to protect light sensitive contents during storage and transport. All Diamond® RealSeal™ amber bottles comply with US Pharmacopoeia 23661 requirements for maximum light transmission.