Diamond® RealSeal™ Bottles, Narrow Mouth, Low Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7070030 7070060 7070125 7070250 7070500 7071000 7072000 7074000 7078000
76360-358PK 37.54 CAD
76360-358 76360-360 76360-362 76360-364 76360-366 76360-368 76360-370 76360-372 76360-374
Diamond® RealSeal™ Bottles, Narrow Mouth, Low Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Better clarity than HDPE
  • Flexible with excellent impact resistance
  • Medical grade resins meet demanding requirements
  • Leakproof, linerless caps for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These narrow mouth bottles improve pouring of liquid contents making them perfect for storing and shipping liquids.