Diamond® RealSeal™ Bottles, Wide Mouth, Low Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7020030 7020060 7020125 7020250 7020500 7021000 7022000
76360-294PK 53.2 CAD
76360-294 76360-296 76360-298 76360-300 76360-302 76360-304 76360-306
Diamond® RealSeal™ Bottles, Wide Mouth, Low Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Better clarity than HDPE
  • Flexible with excellent impact resistance
  • Medical grade resins meet demanding requirements
  • Leakproof, linerless caps for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant
More Product Information