Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7010030 7010060 7010125 7010250 7010500 7011000 7012000 7174000 7184000
76360-258PK 56.05 CAD
76360-258 76360-260 76360-262 76360-264 76360-266 76360-268 76360-270 76360-272 76360-274
Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles
These are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber, even large format with handles, the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These wide mouth bottles are perfect for sample collection and facilitate rapid filling and emptying of solids, powders, and viscous liquids.