Empore™ Sealing Tape Pads, CDS Analytical
Supplier: CDS Analytical
Empore®
660
76333-160CS 966.58 CAD
76333-160
Empore™ Sealing Tape Pads, CDS Analytical
Seals Microplate Seals
Empore™ sealing tape pads are ideal for covering a 96-well plate during storage or transfer to another work station. They are not intended for use with auto-injectors.
Empore™ tape pads are intended for sealing unused portions of a 96-well solid phase extraction plate during vacuum processing to enable vacuum to be uniformly applied to each well being used. Use a clean sheet each time for maximum sealing ability.
Caution: These tape sheets are not intended for sealing collection plates prior to sample injection because the adhesive may clog the injection needle.