Empore™ Solid Phase Extraction Disks, CDS Analytical
Supplier: CDS ANALYTICAL
98-0405-0062-5 98-0405-0063-3 98-0604-0214-0 98-0604-0215-7 98-0604-0217-3 98-0604-0218-1 98-0604-0223-1 98-0604-0224-9 98-0604-0226-4 98-0604-0229-8 98-0604-0230-6 98-0604-0232-2 98-0604-0235-5 98-0604-0236-3 98-0604-0240-5
76333-102CS 557.9 CAD
76333-102 76333-104 76333-128 76333-130 76333-132 76333-134 76333-136 76333-138 76333-140 76333-142 76333-144 76333-146 76333-148 76333-150 76333-152 76333-154 76333-162
Empore™ Solid Phase Extraction Disks, CDS Analytical
Filters Filter Discs
Empore SPE disks are a cost-effective and trusted solution in large-volume sample processing for more than 30 years.
  • Large-volume processing – Ultra-fast flow up to 700 ml/min can be achieved
  • Avoid fine particulates – Particulates cause clogging on column frits, which is common with glass-fiber disks; with the Empore disk, sorbent is tightly bound in a polymer matrix eliminating the shedding of fine particulates
  • EPA compliant with drinking and wastewater testing methods
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore SPE disks are EPA compliant, have excellent reproducibility, are optimized for fast flow for large volume samples, and do not contribute to degradation of column life due to the unique nature of the Empore membrane.