Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Supplier: CDS ANALYTICAL
98-0405-0064-1 98-0405-0065-8 98-0405-0066-6
76333-106CS 13502.52 CAD
76333-106 76333-108 76333-110
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Filters Filter Discs
Empore RAD disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.
  • EPA compliant with radiochemical testing methods
  • Improved safety by reducing usage of concentrated acids needed for precipitation methods
  • Easier sample handling and waste management by avoiding excessively large sample volumes needed for other radiochemical testing methods
  • Large-volume processing – Ultra-fast flow up to 700 mL/min can be achieved
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore RAD SPE disks trap radiochemical species by molecular recognition technology. Concentration, separation, and sample counting preparation are all easily combined into a single step.