VWR® Ratchet Headgear with Face Shield
Supplier: VWR
VWR®
I-1BG-01 I-1
76336-852CS 917.32 CAD
76336-852 76336-850
VWR® Ratchet Headgear with Face Shield
Face Shields
The VWR® Ratchet Headgear with Face Shield provides full face protection from debris and splash hazards. Ideal for use in many environments ranging from the lab to industrial applications.
  • Lightweight design
  • Ratchet headgear features padded headband for better fit and comfort
  • Face shield features universal slot design compatible with many different headgear styles
  • Fits comfortably with respirator or eyewear
  • Impact-resistant, scratch-resistant polycarbonate shield
Face shields should be worn with ANSI approved eyewear.
Certifications: ANSI Z87.1 certified