Hydrochloric acid 0.1 mol/l (0.1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ200

1.09060.1000 1.09060.5000 1.09060.9025 1.09060.4000 1.09060.9010
CA1.09060.1000CS 264 CAD
CA1.09060.1000 CA1.09060.5000 CA1.09060.9025 CA1.09060.4000 CA1.09060.9010
Hydrochloric acid 0.1 mol/l (0.1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Hydrochloric acid

In North America 1.09060.1000 is identical to HX0603A-8 but in a larger pack size.
In North America 1.09060.1000 is identical to HX0603A-6.
In North America 109060.4000 is identical to HX0603A-3 but in a superior shelf life enhancing pack.


Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: HCl
MW: 36.46
Density: 1.00 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR