Hydrochloric acid 0.5 mol/l (0.5 N), Titripur® Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

1.09058.1000 1.09058.4000 1.09058.9025
CA1.09058.1000CS 272 CAD
CA1.09058.1000 CA1.09058.4000 CA1.09058.9025
Hydrochloric acid 0.5 mol/l (0.5 N), Titripur® Reag. USP volumetric solution, Supelco®
Hydrochloric acid
Formula: HCl
Density: 1.01 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR