Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.09057.9010 1.09057.1000 1.09057.2500 1.09057.5000 1.09057.9025 1.09057.4000 1.09057.9010
EM1.09057.9010EA 276.5 CAD
EM1.09057.9010 CA1.09057.1000 CA1.09057.2500 CA1.09057.5000 CA1.09057.9025 CA1.09057.4000 CA1.09057.9010
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Hydrochloric acid

In North America 1.09057.1000 is identical to HX0603D-8 but in a larger pack size.

Formula: HCl
Density: 1.02 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Amount-of-substance concentration 0.995 - 1.005 mol/L
Form liquid
Measurement uncertainty ± 0.003 mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR