Sulfuric acid 0.25 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

1.09073.1000 1.09073.4000 1.09073.9010
CA1.09073.1000CS 302 CAD
CA1.09073.1000 CA1.09073.4000 CA1.09073.9010
Sulfuric acid 0.25 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR