Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ100
1.09139.1000
1.09139.9010
CA1.09139.1000CS
272
CAD
CA1.09139.1000
CA1.09139.9010
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sodium hydroxide
Formula:
NaOH Boiling Pt: ∼105 °C (1013 hPa) Density: 1.02 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Form | liquid |
Amount-of-substance concentration | 0.24875 - 0.25125 |
Measurement uncertainty | 0.00075 |
Traceability | NIST SRM |
The concentration is determined by volumetric titration and refers to 20°C. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...