Sodium hydroxide 0.5 mol/l (0.5 N) aqueous solution, Titripur® Volumetric, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Caustic soda

MQ100

1.09138.1000 1.09138.4000 1.09138.9010
CA1.09138.1000CS 276 CAD
CA1.09138.1000 CA1.09138.4000 CA1.09138.9010
Sodium hydroxide 0.5 mol/l (0.5 N) aqueous solution, Titripur® Volumetric, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025
Measurement uncertainty +/- 0.0015
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR