Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.09137.1000 1.09137.2500 1.09137.9025 1.09137.4000 1.09137.9010
CA1.09137.1000CS 284 CAD
CA1.09137.1000 CA1.09137.2500 CA1.09137.9025 CA1.09137.4000 CA1.09137.9010
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Sodium hydroxide

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: NaOH
Density: 1.04 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.995 - 1.005
Measurement uncertainty +/- 0.003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR