Sodium hydroxide 2 mol/l (2 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda

M-Clarity™ quality level = MQ100

1.09136.1000 1.09136.9025
CA1.09136.1000CS 289 CAD
CA1.09136.1000 CA1.09136.9025
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sodium hydroxide
Formula: NaOH
Density: 1.09 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 1.990 - 2.010
Measurement uncertainty +/- 0.006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR