Perimeter Workstation, Diversified Woodcrafts
Supplier: Diversified Woodcrafts
2816KF
76292-578EA 12832.08 CAD
76292-578
Perimeter Workstation, Diversified Woodcrafts
Furniture Tables and Workbenches
This perimeter workstation provides work space for up to four students and valuable storage space. Made of solid oak, oak and hardwood veneers.
  • 1" solid epoxy resin surface
  • Finished with rubber base molding and a durable chemical resistant UV finish
  • Flat work surface for maximum workspace
  • Full cabinet doors with locks
  • Two GFI protected AC duplex receptacles
Provides work space for up to four-students while storing your laboratory equipment in these perimeter workstations. The four cabinets which make up a single unit measure 24W×16D×35"H. The stations are designed to utilize space along walls where they can stand alone or be ganged together. Assembly and installation of electricity and plumbing is required.
Sink measures: 12W×8D×6"H. Work surface on wall portion is 18"D.
Certifications: SEFA compliant and MAS Certified Green.
Delivery information: Each unit includes: 1" epoxy resin with back splash, solid epoxy drop-in style sink with strainer, 2 combination cold water and gas fixtures, 4 rod sockets, rod storage (rods not included), 2 GFI electrical receptacles, keyed-alike locks on all doors and dovetailed drawers and rubber base molding.