Backpack Cabinets, Diversified Woodcrafts
Supplier: Diversified Woodcrafts
BP-1215-51K
BP-1215-51M
BP-1215-72K
BP-1215-72M
BP-2415-51K
BP-2415-51M
BP-2415-72K
BP-2415-72M
BP-3615-51K
BP-3615-51M
BP-3615-72K
BP-3615-72M
BP-4815-51K
BP-4815-51M
BP-4815-72K
BP-4815-72M
76292-592EA
1381.71
CAD
76292-592
76292-594
76292-596
76292-598
76292-600
76292-602
76292-604
76292-606
76292-608
76292-616
76292-618
76292-620
76292-622
76292-624
76292-626
76292-628
Backpack Cabinets, Diversified Woodcrafts
Furniture
Storage Cabinets
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...