Long-Handled Water Dippers
Supplier: Bel-Art Products
F36785-0016 F36784-0016
470323-316EA 146.4 CAD
470323-316 470323-318
Long-Handled Water Dippers
Samplers Water Samplers
Ideal for sampling ponds and lakes.
  • Two pouring spouts on cup allow left or right pouring
  • High-density polyethylene plastic handles are strong honeycomb tubes without inserts
  • Handles are ribbed for a sure grip
  • Handle end has a hook for hanging storage
Collect water samples while remaining on the shore; the long handles of the water dippers let you reach three or six feet from dry land. Lightweight and easy to use, each dipper is made of durable plastic to withstand years of use. The molded 16 oz. cup screws onto the handle and has two pouring spoutsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable