VWR® Book Compartment Tables with Cabinet for Two Student
Supplier: VWR International
89374-858 89374-874 89374-880 89374-882
89374-858EA 6572.64 CAD
89374-858 89374-874 89374-880 89374-882
VWR® Book Compartment Tables with Cabinet for Two Student
Furniture Tables and Workbenches
Two student cabinet table constructed of solid oak, oak and hardwood veneers.
  • 1" solid epoxy resin surface
  • Includes two book compartments in the aprons
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
  • Two student workstation features a locking center cabinet with two locking drawers
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plainMore Product Information